Anti-IgG Goat Polyclonal Antibody (Texas Red®)
Catalog # CA601-1902
Supplier: Rockland Immunochemical
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Texas Red®
- Flow cytometry:Yes
- Host:Goat
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Bovine
- Size:2 mg
- Epitope:Heavy and Light chain
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide, 10 mg/ml Polyethylene Glycol (PEG-8000)
- Storage temperature:–20 °C
- Concentration:2.0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ambient
- Immunogen:Bovine IgG whole molecule
- Purification:purified by immunoaffinity chromatography
- Cat. no.:CA601-1902
- Supplier No.:601-1902
Specifications
About this item
Secondary Goat Anti-IgG (H&L) Reacts with Cow (Bovine, Cattle)
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Bovine