To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Bovine
- Western blot:Yes
- Size:2 mL
- Epitope:Heavy and Light chain
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Storage buffer:0.01 M Sodium Phosphate, 0.25 M Sodium Chloride, pH 7.2. 15 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
- Storage temperature:–20 °C
- Concentration:0.8 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ambient
- Immunogen:Bovine IgG whole molecule
- Purification:purified by immunoaffinity chromatography
- Cat. no.:CA601-1302
- Supplier No.:601-1302
Specifications
About this item
Anti-Bovine IgG (heavy and light chain) Antibody is peroxidase conjugated and is suitable for immunoassays where specificity to bovine immunoglobulin is desired. Antibody has been tested by ELISA, western blot immunoblot, and immunohistochemisty.
Recommended Dilutions: ELISA: 1:10000 - 1:200,000; Western Blot: 1:5000 - 1:100000; Immunochemistry: 1:500 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Bovine