Anti-IgG1 gamma Rabbit Polyclonal Antibody (Biotin)
Catalogue # CA610-4640
Fournisseur: Rockland Immunochemical
Spécifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G1 gamma
- Antigen symbol:IgG1 gamma
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Size:1 mg
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
- Storage temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ambient
- Immunogen:Mouse IgG1 heavy chain
- Purification:purified by immunoaffinity chromatography
- Cat. no.:CA610-4640
- Supplier No.:610-4640
Spécifications
À propos de cet article
Secondary Rabbit Anti- IgG1 Reacts with Mouse
Recommended Dilutions: ELISA: 1:250000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:3000;
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgG1 gamma
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse