Order Entry
Canada
ContactUsLinkComponent
Anti-GALM Rabbit Polyclonal Antibody [clone: C11]
Catalog # CA100-4162
Anti-GALM Rabbit Polyclonal Antibody [clone: C11]
Catalog # CA100-4162
Supplier Number:  100-4162
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Galactose mutarotase
  • Antigen symbol:
    GALM
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Pig
  • Western blot:
    Yes
  • Size:
    2 mL
  • Cross adsorption:
    No
  • Format:
    Antiserum
  • Form:
    Lyophilized
  • Gene ID:
    399536
  • Antigen synonyms:
    Galactose mutarotase|BLOCK25 protein|GALM|Aldose 1 epimerase
  • Storage buffer:
    0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
  • Storage temperature:
    –20 °C
  • Concentration:
    90 mg/ml by Refractometry
  • Shipping temperature:
    Ambient
  • Immunogen:
    Mutarotase (Porcine Kidney)
  • Purification:
    delipidation and defibrination
  • Cat. no.:
    CA100-4162
  • Supplier No.:
    100-4162

Specifications

About this item

This product has been assayed against 1.0 µg of Mutarotase (Porcine Kidney) in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG (H&L) (Goat) and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: GALM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Pig