To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Interleukin-1 Receptor-associated Kinase 1
- Antigen symbol:IRAK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Antigen synonyms:IRAK|IL-1 Receceptor Associated Kinase|pelle|mPLK|Pelle|Pelle homolog
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Dry Ice
- Immunogen:This IgG fraction antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a region near the carboxy terminus of human IRAK Protein.
- Purification:purified by DEAE chromatography
- Cat. no.:CARL200401-951
- Supplier No.:200-401-951
Spécifications
À propos de cet article
This IgG fraction antibody has been tested for use in ELISA, immunoprecipitation, immunochemistry and western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 80 kDa in size.
Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:100 - 1:500; Immunoprecipitation: 2-4 µg per sample;
Type: Primary
Antigen: IRAK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat