Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Anti-IgG Rabbit Polyclonal Antibody | Avantor

Anti-IgG Rabbit Polyclonal Antibody

2 mL
Catalog # CA108-4104
Fournisseur #:  108-4104
Anti-IgG Rabbit Polyclonal Antibody
Catalog # CA108-4104
Anti-IgG Rabbit Polyclonal Antibody

2 mL

Price
$301.69
Log in pour la disponibilité
Each (2ml)
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Secondary Rabbit Anti-IgG F(ab')2 Reacts with Horse (Equine)

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Horse

Spécifications
  • Taille:
    2 mL
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Purification:
    delipidation and defibrination
  • Antibody type:
    Secondary
  • Epitope:
    F(ab')2
  • Form:
    Lyophilized
  • Isotype:
    IgG
  • Storage temperature:
    –20 °C
  • Host:
    Rabbit
  • Concentration:
    31.0 mg/ml by Refractometry
  • Cross adsorption:
    No
  • Shipping temperature:
    Ambient
  • Antigen name:
    Immunoglobulin G
  • Cat. No.:
    CA108-4104
  • Reactivity:
    Horse
  • Format:
    Antiserum
  • Immunogen:
    Horse IgG F(ab')2 fragment
  • ImmunoChemistry:
    Yes
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    IgG
  • Storage buffer:
    0.01 M Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2,
  • ELISA:
    Yes
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.