Order Entry
Canada
ContactUsLinkComponent
Anti-IgM µ Rabbit Polyclonal Antibody
  CA104-4107
Anti-IgM µ Rabbit Polyclonal Antibody
  CA104-4107
 :  104-4107
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin M mu-chain
  • Antigen symbol:
    IgM µ
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgM
  • Reactivity:
    Dog
  • Western blot:
    Yes
  • Size:
    2 mL
  • Epitope:
    µ-chain
  • Cross adsorption:
    No
  • Format:
    Antiserum
  • Form:
    Lyophilized
  • Storage buffer:
    0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
  • Storage temperature:
    –20 °C
  • Concentration:
    75 mg/ml by Refractometry
  • Shipping temperature:
    Ambient
  • Immunogen:
    Dog IgM mu heavy chain
  • Purification:
    delipidation and defibrination
  • Cat. no.:
    CA104-4107
  • Supplier No.:
    104-4107

 

 

Secondary Rabbit Anti-IgM mu chain Reacts with Dog (Canine)

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: µ-chain
Host: Rabbit
Isotype: IgM
Reactivity: Dog