Order Entry
Canada
ContactUsLinkComponent
Anti-IgG3 gamma Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalogue # CA610-4543
Anti-IgG3 gamma Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalogue # CA610-4543
Supplier Number:  610-4543

Spécifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G3 gamma
  • Antigen symbol:
    IgG3 gamma
  • Clonality:
    Polyclonal
  • Conjugation:
    AP (Alkaline Phosphatase)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Size:
    1 mg
  • Cross adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Storage buffer:
    0.05 M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0, 0.01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
  • Storage temperature:
    +4 °C
  • Concentration:
    1.0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Wet Ice
  • Immunogen:
    Mouse IgG3 heavy chain
  • Purification:
    purified by immunoaffinity chromatography
  • Cat. no.:
    CA610-4543
  • Supplier No.:
    610-4543

Spécifications

À propos de cet article

This product has been assayed against 1.0 µg of Mouse IgG in a standard capture ELISA using pNPP p-nitrophenyl phosphate as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgG3 gamma
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse