Anti-IgM µ Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
CA618-105-007
- Antibody type:Secondary
- Antigen name:Immunoglobulin M mu-chain
- Antigen symbol:IgM µ
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgM
- Reactivity:Ferret
- Western blot:Yes
- Size:1 mg
- Epitope:µ-chain
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Storage buffer:0.05 M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0, 0.01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
- Storage temperature:+4 °C
- Concentration:1.1 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Wet Ice
- Immunogen:Ferret IgM mu heavy chain
- Purification:purified by immunoaffinity chromatography
- Cat. no.:CA618-105-007
- Supplier No.:618-105-007
Secondary Goat Anti-IgM (mu chain) Reacts with Ferret
Recommended Dilutions: ELISA: 1:1000 - 1:2500; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!
Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: µ-chain
Host: Goat
Isotype: IgM
Reactivity: Ferret