Anti-PDCD4 Rabbit Polyclonal Antibody
100 µg
Catalog # CA600-401-965
Supplier: Rockland Immunochemical
Fournisseur #: 600-401-965
Jump to:
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
This affinity purified antibody has been tested for use in ELISA, western blotting, immunoprecipitation and immunochemistry. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 52 kDa in size.
Recommended Dilutions: ELISA: 1:10000 - 1:120000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 1:100 – 1:300;
Type: Primary
Antigen: PDCD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Xenopus
Spécifications
- Taille:100 µg
- Western blot:Yes
- Clonality:Polyclonal
- ImmunoPrecipitation:Yes
- Purification:affinity-purified
- Antibody type:Primary
- Form:Liquid (sterile filtered)
- Isotype:IgG
- Storage temperature:–20 °C
- Host:Rabbit
- Concentration:1.06 mg/ml by UV absorbance at 280 nm
- Cross adsorption:No
- Shipping temperature:Dry Ice
- Antigen name:Programmed cell death protein 4
- Cat. No.:CA600-401-965
- Reactivity:Xenopus|Human|Rat|Mouse
- Format:Antibody fraction
- Gene ID:27250
- Antigen synonyms:Ma3|Neoplastic transformation inhibitor protein|Nuclear antigen H731|H731|Neoplastic transformation inhibitor|Dug|Death up-regulated gene protein
- Immunogen:This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding amino acids near the carboxyl terminus of human Pdcd4 Protein.
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:PDCD4
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- ELISA:Yes



