Order Entry
Canada
ContactUsLinkComponent
Anti-CBX4 Rabbit Polyclonal Antibody
Catalogue # CARL600401-478
undefined
Anti-CBX4 Rabbit Polyclonal Antibody
Catalogue # CARL600401-478
Supplier Number:  600-401-478
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Spécifications

  • Antibody type:
    Primary
  • Antigen name:
    Chromobox Homolog 4
  • Antigen symbol:
    CBX4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Cross adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Antigen synonyms:
    Chromobox protein homolog 4|E3 SUMO-protein ligase CBX4|PC2|NBP16|Pc2|CBX4
  • Amino acid number:
    95 to 107
  • Storage buffer:
    0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
  • Storage temperature:
    –20 °C
  • Concentration:
    1.36 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Dry Ice
  • Immunogen:
    This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to aa 95-107 of Human PC2 Protein.
  • Purification:
    affinity-purified
  • Cat. no.:
    CARL600401-478
  • Supplier No.:
    600-401-478

Spécifications

À propos de cet article

This affinity purified antibody has been tested for use in immunofluorescence staining of cultured cells and western blotting using transfected cell lysates.  The antibody is also functional by ELISA.  Reactivity in other immunoassays is unknown.

Recommended Dilutions: ELISA: 1:10000 - 1:40000; Immunofluorescence: 1:100 - 1:400; Western Blot: 1:1000 - 1:4000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CBX4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human