Order Entry
Canada
ContactUsLinkComponent
Anti-IgG Rabbit Polyclonal Antibody
Anti-IgG Rabbit Polyclonal Antibody
Catalog # CA607-4104
Anti-IgG Rabbit Polyclonal Antibody
Catalog # CA607-4104
Supplier Number:  607-4104
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Golden syrian hamster
  • Western blot:
    Yes
  • Size:
    5 mg
  • Epitope:
    F(ab')2
  • Cross adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Storage buffer:
    0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
  • Storage temperature:
    –20 °C
  • Concentration:
    5.0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Wet Ice
  • Immunogen:
    Hamster IgG F(ab')2 fragment
  • Purification:
    purified by immunoaffinity chromatography
  • Cat. no.:
    CA607-4104
  • Supplier No.:
    607-4104

Specifications

About this item

Secondary Rabbit Anti-IgG F(ab')2 Reacts with Hamster

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Golden Syrian Hamster