Order Entry
Canada
ContactUsLinkComponent
Anti-PCNA Rabbit Polyclonal Antibody
Catalogue # CA600-401-887
undefined
Anti-PCNA Rabbit Polyclonal Antibody
Catalogue # CA600-401-887
Supplier Number:  600-401-887
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Spécifications

  • Antibody type:
    Primary
  • Antigen name:
    Proliferating cell nuclear antigen
  • Antigen symbol:
    PCNA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Isochrysis galbana,
    Algae
  • Western blot:
    Yes
  • Size:
    100 µg
  • Cross adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Antigen synonyms:
    DNA polymerase delta auxiliary protein|HGCN8729|MGC8367|Proliferating Cell Nuclear Antigen|cyclin|polymerase delta accessory protein
  • Amino acid number:
    55 to 71
  • Storage buffer:
    0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
  • Storage temperature:
    –20 °C
  • Concentration:
    1.42 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Dry Ice
  • Immunogen:
    This affinity-purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 55-71 of Isochrysis galbana PCNA.
  • Purification:
    affinity-purified
  • Cat. no.:
    CA600-401-887
  • Supplier No.:
    600-401-887

Spécifications

À propos de cet article

This affinity purified antibody has been tested for use in ELISA and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band at ~29 kDa in size corresponding to PCNA by western blotting in the appropriate cell lysate.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:5000 - 1:32000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: PCNA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Algal, Isochrysis galbana