To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Description:PBS, Phosphate Buffered Saline [10X]
- Size:500 mL
- Cat. no.:CA89167-714
- Supplier No.:786-027
Spécifications
À propos de cet article
G-Biosciences' Phosphate and Tris buffered saline solutions are both conveniently supplied in a 10X concentrated, ready-to-use format.
G-Biosciences' Tris Buffered Saline (TBS) contains one to five percent 100mM Tris HCl (CAS# 77-86-1) and less than ten percent 1.5M sodium chloride (CAS# 7647-14-5). The solution is at a pH of 7.5.
G-Biosciences' Phosphate BufferedSane (PBS) contains 1.5M sodium chloride (CAS# 7647-14-5), 30mM potassium chloride (CAS# 7447-40-7), 80mM sodium phosphate monobasic, and 20mM potassium phosphate dibasic. The solution is at a pH of 7.4.
G-Biosciences also offers convenient, easy-to-use, pre-mixed dry powder buffer packs that simply require adding water to generate for use. G-Biosciences’ JAW™ buffered saline packs are offered in the following formats: Phosphate buffered saline packs (available as either 1X or 10X concentrated), Carbonate-Bicarbonate buffered packs (1X concentrated), or Tris buffered saline packs (available as either 1X or 20X concentrated).