ContactUsLinkComponent
Fluorescent β-Galactosidase Assay, G-Biosciences
Catalogue # CA89167-842
Fournisseur:  G-Biosciences
Numéro CAS:  
undefined
Fluorescent β-Galactosidase Assay, G-Biosciences
Catalogue # CA89167-842
Fournisseur:  G-Biosciences
Supplier Number:  786-654
Numéro CAS:  
Spécifications
  • Description:
    β-Galactosidase Assay (CPRG)
  • Size:
    500 assays
  • Cat. no.:
    CA89167-842
  • Supplier No.:
    786-654
Spécifications
À propos de cet article
G-Biosciences' Fluorescent β-Galactosidase Assay (MUG) is a highly sensitive, fluorescent assay for determining the β-galactosidase activity in the lysates of cells transfected with a β-galactosidase expression construct.
The study of the lac operon has played an important role in understanding the control of gene expression in bacteria. In prokaryotes, gene expression is controlled primarily at the level of transcription. For eukaryotes, the promoter activity can be analyzed by using fusion genes containing the promoter of interest attached to the bacterial β-galactosidase gene and be assayed by measuring β-galactosidase activity. Because β-galactosidase has a high turnover rate and is absent in mammalian cells it serves as a very useful and sensitive reporting tool for gene expression.
Esters of the fluorescent compound, 4-methylumbelliferone (4-MU), provide a sensitive, quantitative assay for β-galactosidase. 4-methylumbelliferyl-β-D-galactopyranoside (4-MUG) is a substrate of β-galactosidase that does not fluoresce until cleaved by the enzyme to generate the fluorophore 4-methylumbelliferone. The assay can be used with extracts from different expression systems including mammalian, insect cells, yeast, and bacteria.
G-Biosciences' Fluorescent β-Galactosidase Assay (MUG) provides a 96 well assay format for galactosidase activity that is suitable for high throughput applications. The production of the fluorphore is monitored at an emission/excitation wavelength of 365/460nm. This kit is designed for 500 micro-well assays.
Ordering information: Accessories available