ContactUsLinkComponent
CytoScan™-fluoro Cytotoxicity Assay, G-Biosciences
CytoScan™-fluoro Cytotoxicity Assay, G-Biosciences
Catalog # CA82021-584
Supplier:  G-Biosciences
CAS Number:  
undefined
CytoScan™-fluoro Cytotoxicity Assay, G-Biosciences
Catalog # CA82021-584
Supplier:  G-Biosciences
Supplier Number:  786-211
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    CytoScan™-fluoro Assay Kit
  • Size:
    500 assays
  • Cat. no.:
    CA82021-584
  • Supplier No.:
    786-211
Specifications
About this item
G-Biosciences' CytoScan™-fluoro Cytotoxicity Assay, for the detection of cell toxicity, death, viability or proliferation, is ideal for high throughput screening.
G-Biosciences' CytoScan™-fluoro Cytotoxicity Assay is based on the quantification of cellular lactate dehydrogenase (LDH) released when cells are damaged or under stress. Lactate dehydrogenase released into the culture medium is measured with a diaphorase coupled enzymatic assay that results in the conversion of a non-fluorescent compound (resofurin) to a fluorescent compound (resorufin), which is measured using a fluorometer. The assay detects even low level damage to cell membrane not detected with other methods.
G-Biosciences' CytoScan™-fluoro Cytotoxicity Assay does not damage healthy cells and the assay can be performed directly in the cell culture wells containing a mixed population of viable and damaged cells. The kit is supplied with substrate mix, assay buffer, and stop solution.
G-Biosciences' CytoScan™-fluoro Cytotoxicity Assay is supplied in a kit that contains components sufficient for performing up to 500 assays.
Ordering information: Accessories available