Botanical Garden Blot, G-Biosciences
CA82022-768
- Type:—
- Description:Botanical Garden Blot
- Blot lane order:1. Protein marker
2. Green Algae Lysate
3. Yeast Lysate
4. Barley Lysate
5. Sweet Corn Lysate
6. Rye Grass Lysate
7. Oat Lysate
8. Wheat Lysate
9. Soy Lysate
10. Mung Lysate
11. Sunflower Lysate
12. Cotton Lysate
13. Beet Lysate
14. Tomato Lysate
15. Spinach Lysate - Cat. no.:CA82022-768
- Host:Botanical
- Source:Various
- Tissue:Multiple Species
- Supplier No.:BB01
G-Biosciences' Botanical Garden Blot is prepared using total protein extracted from a variety of 5-10 day old, whole plant tissues.
G-Biosciences' premade blots are prepared with the appropriate GenLysate™ and 50 μg protein is loaded onto a 4-20% denaturing polyacrylamide gel, along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane, using our Efficient Western Transfer Buffer.
G-Biosciences' premade blots are ready to be blocked and probed with the antibodies of choice. Note: We recommend that customers contact our technical department for the latest information on the blots as G-Biosciences reserves the right to change the blot profile due to the availability of GenLysate™.
: Accessories available.