Human Normal Cell Line Blots, G-Biosciences
Supplier: G-Biosciences
Certificates
About this item
G-Biosciences offers ready-to-screen, normal tissue human cell line blots from epithelial, endothelial, skin, and muscle tissue.
G-Biosciences' premade blots are prepared with the appropriate GenLysate™ and 50 μg protein is loaded onto a 4-20% denaturing polyacrylamide gel, along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane, using our Efficient Western Transfer Buffer.
G-Biosciences' premade blots are ready to be blocked and probed with the antibodies of choice. Note: We recommend that customers contact our technical department for the latest information on the blots as G-Biosciences reserves the right to change the blot profile due to the availability of GenLysate™.
Ordering information: Accessories available.
Specifications
- Catalog No:
- CA82023-236
- CA82023-232
- CA82023-234
- CA82023-230
- Blot lane order:
- 1. Protein marker
2. Aortic Smooth Muscle Cells (AoSMC)
3. Bronchial/Tracheal Smooth Muscle Cells (BSMC)
4. Coronary Artery Smooth Muscle Cells (CASMC)
5. Pulmonary Artery Smooth Muscle Cells (PASMC)
6. Umbilical Artery Smooth Muscle Cells (UASMC)
7. Uterine Smooth Muscle Cells (UtSMC)
8. Skeletal Muscle Cells (SKMC) - 1. Protein marker
2. Pulmonary Artery Endothelial Cells (HPAEC)
3. Coronary Artery Endothelial Cells (HCAEC)
4. lliac Artery Endothelial Cells (HIAEC)
5. Aortic Endothelial Cells (HAEC)
6. Lung Microvascular Endothelial Cells (HMVEC-L)
7. Umbilical Vein Endothelial Cells (HMVEC)
8. Umbilical Artery Endothelial Cells (HUAEC)
9. Dermal Microvascular Endothelial Cells (HMVEC-d Ad) - 1. Protein marker
2. Epidermal Keratinocytes Adult (NHEK-Ad0
3. Epidermal Keratinocytes NEO (NHEK-Neo)
4. Epidermal Keratinocytes Neo Pool (NHEK-Neo Pool)
5. Dermal Fibroblast Adult (NHDF-Ad)
6. Dermal Fibroblast Neo (NHDF-Neo)
7. Microvascular Endothelial Adult (HMVECd-Ad)
8. Microvascular Endothelial Neo (HMVECd-Neo) - 1. Protein marker
2. Human Normal Mammary Epithelial Cells (HMEC)
3. Human Normal Renal Cortical Epithelial Tissue (HRCE)
4. Renal Proximal Tubule Epithelial Cells (RPTEC)
5. Bronchial Epithelial cells (NHBE)
6. Prostate Epithelial cells (PrEC)