ContactUsLinkComponent
Multiple Tissue Single Species Blot, G-Biosciences
Multiple Tissue Single Species Blot, G-Biosciences
Supplier:  G-Biosciences
Certificates
undefined
Multiple Tissue Single Species Blot, G-Biosciences
Supplier:  G-Biosciences
Specifications
  • Catalog No:
  • CA82023-206
  • CA82023-208
  • Blot lane order:
  • 1. Protein marker
    2. Mouse Normal Liver Lysate
    3. Mouse Normal Brain Lysate
    4. Mouse Normal Lung Lysate
    5. Mouse Normal Kidney Lysate
    6. Mouse Normal Spleen Lysate
    7. Mouse Normal Testis Lysate
    8. Mouse Normal Ovary Lysate
    9. Mouse Normal Heart Lysate
    10. Mouse Normal Pancreas Lysate
  • 1. Protein marker
    2. Rat Normal Liver Lysate
    3. Rat Normal Brain Lysate
    4. Rat Normal Lung Lysate
    5. Rat Normal Kidney Lysate
    6. Rat Normal Spleen Lysate
    7. Rat Normal Testis Lysate
    8. Rat Normal Ovary Lysate
    9. Rat Normal Heart Lysate
    10. Rat Normal Pancreas Lysate