ContactUsLinkComponent
Nalgene® Benchtop Beta Waste Container (with bottle); Acrylic, PE bottle, Thermo Scientific
Nalgene® Benchtop Beta Waste Container (with bottle); Acrylic, PE bottle, Thermo Scientific
Catalog # 56616-035
undefined
Nalgene® Benchtop Beta Waste Container (with bottle); Acrylic, PE bottle, Thermo Scientific
Catalog # 56616-035
Supplier Number:  6710-2000
Specifications
  • Ø:
    14 cm (51/2")
  • Capacity:
    2 L (64 oz.)
  • Cat. no.:
    56616-035
  • Supplier No.:
    6710-2000
Specifications
About this item
Nalgene Benchtop Beta Waste Container is constructed of 0.95 cm (3/8") thick acrylic
  • Constructed of 0.95 cm (3/8") thick acrylic
  • Cylindrical shield protects personnel from waste material
Provided with a 2-liter HDPE Nalgene wide mouth waste bottle for holding liquid wastes. Transparent shield encases the waste bottle without obstructing access to or manipulation of bottle closure. Machined lip on the cap prevents slipping and provides extra impact protection.
The cylindrical shield protects personnel from waste material contaminating beta-emitting isotopes.