Nalgene® Safety Splash Shield, PC, Acetal Flexible Arm, Magnetic Base, Thermo Scientific
Catalog # 28196-342
Supplier: Thermo Fisher Scientific
Specifications
- Description:Safety splash shield
- Dimensions:28Lx 21.5W cm (12x81/2")
- Cat. no.:28196-342
- Supplier No.:6356-0001
Specifications
About this item
Nalgene Safety Splash Shield protects the user while simultaneously providing a clear field of vision
- Protects from splashes, aerosols and flying debris
- Clear field of vision
- Attaches easily to benchtop shelving, machinery or other magnetic surfaces
- Flexible arm is easy to maneuver
Attaches easily to benchtop shelving, machinery or other magnetic surfaces for convenient protection from splashes, aerosols and flying debris. Flexible arm is easy to maneuver.