Nalgene® Diagnostic Bottles, Natural PP, with Closures, Tray Packed, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Nalgene natural polypropylene diagnostic bottles with natural polypropylene closures are used in test kits and many small volume applications.
Diagnostic bottles can be used for sample storage and shipping of small volume, valuable reagents.
Specifications
- Catalog No:
- 16059-690
- 16059-682
- 16059-698
- Environmentally Preferable:
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)