ContactUsLinkComponent
Nalgene® L900 Liquid Detergent, Thermo Scientific
Catalog # 28196-320
undefined
Nalgene® L900 Liquid Detergent, Thermo Scientific
Catalog # 28196-320
Supplier Number:  900-4000
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    4 L (1 gal.)
  • Cat. no.:
    28196-320
  • Supplier No.:
    900-4000
Specifications
About this item
Nalgene L900 Liquid Detergent is formulated to thoroughly clean all plastic labware, especially polycarbonate
The detergent will not cause crazing, stress cracking or clouding when used as directed. This liquid detergent are for hand or machine use, it is biodegradable, not toxic, and low foaming.