Arrest™ Extraction Buffer, A Chaotropic RNA Extraction Buffer, G-Biosciences
G-Biosciences' Arrest™ Extraction Buffer is an optimized combination of various chaotropic agents and RNase inhibitors, which inhibits RNase in 5-10 minutes
G-Biosciences' Arrest™ Extraction Buffer has a quick, 10 minute, single step protocol that isolates high quality total RNA from most species and tissues. The protocol involves homogenization and lysis of samples in Arrest™ Extraction Buffer. After removing cellular debris, pure RNA is precipitated from the supernatant with ethanol.
G-Biosciences' Arrest™ Extraction Buffer is conveniently packaged as two 50 mL bottles; enough reagent for isolation of RNA from 10 grams of tissue or 200 preps of 50 mg tissue each.
Arrest™ Extraction Buffer may be used in conjunction with any RNA extraction method, including extractions based on phenol, chloroform and other organic solvents and detergents.
: Accessories available.
- Désignation:Arrest™ Extraction Buffer
- Taille:200 preps