GBiosciences' Toothpick™ systems allow for the rapid screening of bacterial colonies by restriction enzyme digestion analysis or polymerase chain reaction (PCR), with our Plasmid Screening Toothpick™ and Toothpick™PCR, respectively.
GBiosciences' Toothpick™ systems eliminate the need for growing overnight cultures, performing minipreps, and purifying or isolating the plasmid DNA, thus producing much faster results. G-Biosciences’ simple, 5 minute protocol is as easy as picking a bacterial colony and adding it to our Toothpick™ solution.
An added advantage of using our Toothpick™PCR is that the supplied Glow™ Loading Dye minimizes the need to stain with ethidium bromide, therefore reducing exposure and the many problems associated with its use and disposal.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Method / Format:Extraction for direct PCR
- Prep size:Scalable
- Refine target molecule:Plasmid
- Sample type:Bacteria
- Target molecule:Plasmid
- Description:Plasmid Screening Toothpick* System