Nalgene® Replacement Coupling Inserts for Quick Filling Venting Closure; PP, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Nalgene Replacement coupling inserts support one handed connection and disconnection when applied to mating couple bodies
They promote fewer leakpoints, short assemblies, and faster installations.
Frequently Bought Together