ContactUsLinkComponent
MegaLong™ for High Molecular Weight DNA Isolation, G-Biosciences
MegaLong™ for High Molecular Weight DNA Isolation, G-Biosciences
Supplier:  G-Biosciences
Certificates
undefined
MegaLong™ for High Molecular Weight DNA Isolation, G-Biosciences
Supplier:  G-Biosciences
Specifications
  • Method / Format:
    Enzymatic extraction
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Sample type:
    Plant samples
    Bacteria
    Yeast/Fungi
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    DNA
  • Description:
    MegaLong™