Corning® Lambda™ Plus Starter Kit, Corning
Lambda Plus Starter Kit
Catalog # 89187-013
Supplier: Corning
Fournisseur #: 4069
Corning® Lambda™ Plus Starter Kit, Corning
Catalog # 89187-013
Supplier: Corning
Jump to:
Product Details & Documents
The Lambda plus starter kit is designed for laboratories which are about to replace their old set of pipettors, and for customers who want to get a new set of pipettors which cover the most commonly used volumes.
The Lambda plus starter kit contains four Lambda Plus Single-Channel pipettors conveniently packaged with useful accessories (manual, calibration key, color-coded rings, tube of piston grease and identification stickers).
Ordering information: This starter kit includes four Lambda Plus Single Channel pipettors in these volume sizes 0.5 to 10 µl, 2.0 to 20 µl, 20 to 200 µl, and 100 to 1000 µl and three racks of Corning IsoTip™ filtered pipette tips for the above sizes.
Spécifications
- Désignation:Lambda Plus Starter Kit
- Cat. No.:89187-013
- Environmental certification:I-RECs, ISO 14001 - Warsaw
- Environmentally Preferable:Low Manufacturing Impact