Mobile Equipment Table, Labconco® | Avantor
Mobile Equipment Table, Labconco®
Supplier:   Labconco
Mobile Equipment Table, Labconco®
Supplier:  Labconco
Certificates
Mobile Equipment Table, Labconco®
Mobile Equipment Table, Labconco®
3 Options Available Below
Product Details & Documents
Mobile equipment table may be used to transport heavy equipment and to provide auxiliary bench space.
  • 5" diameter non-marking polyurethane casters with toe locks have bearings for quiet operation
  • Supports loads up to 800 pounds
With height adjustments from 28.75" to 35.75", it may be raised to the same height as most laboratory countertops. Solid epoxy 1.25" thick × 30" deep work surface and epoxy-coated 1.75" square tubular steel frame.
Product Family Options
Informations sur le produit
  • Désignation
    Hauteur
    W×D
    Availability
    Price
  • Mobile equipment table
    28¼ - 35¼"
    48×30"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,060.00
    $3,060.00
    Each
       $3,060.00
    La quantité minimale de commande est de 1
    Supplier Number: 3819100
  • Mobile equipment table
    28¼ - 34¼"
    60×30"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,520.00
    $3,520.00
    Each
       $3,520.00
    La quantité minimale de commande est de 1
    Supplier Number: 3819101
  • Mobile equipment table
    28¼ - 34¼"
    72×30"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,900.00
    $3,900.00
    Each
       $3,900.00
    La quantité minimale de commande est de 1
    Supplier Number: 3819102