Acid Resistant Refrigerated CentriVap Centrifugal Vacuum Concentrators, Labconco®
Catalogue # 89187-284
Fournisseur: Labconco
Product available on GSA Advantage®
Spécifications
- Description:Acid Resistant Refrigerated CentriVap Centrifugal Vacuum Concentrator
- Cat. no.:89187-284
- Electrical:115 V
- Supplier No.:7310022
Spécifications
À propos de cet article
The acid-resistant refrigerated CentriVap centrifugal concentrator is designed to rapidly concentrate multiple small heat-sensitive samples, such as RNA and proteins, using centrifugal force, vacuum and controlled temperature.
This exclusive concentrator combines a refrigeration system that cools to –4 °C with a heater that warms to 100 °C to achieve the users desired set point temperatures. It also features a corrosion-resistant, Teflon-coated chamber and Hastelloy shaft to protect against samples containing corrosive solvents.