ContactUsLinkComponent
BLOT-FastStain™, G-Biosciences
Catalogue # CA82021-410
Fournisseur:  G-Biosciences
Numéro CAS:  
undefined
BLOT-FastStain™, G-Biosciences
Catalogue # CA82021-410
Fournisseur:  G-Biosciences
Supplier Number:  786-34
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    BLOT-FastStain™
  • Size:
    25 blots
  • Cat. no.:
    CA82021-410
  • Supplier No.:
    786-34
Spécifications
À propos de cet article
BLOT-FastStain™ is a unique stain for reversible staining of protein on nitrocellulose and PVDF transfer membranes. It only stains protein and leaves the background absolutely untouched and brilliant white resulting in high band visibility.
  • Rapid and sensitive staining of proteins on Western blots.
  • Reversible stain for protein membranes
  • Compatible with PVDF and nitrocellulose membranes
  • Detect >2ng protein
BLOT-FastStain™ only stains protein and leaves the background absolutely untouched and brilliant white resulting in high band visibility. The staining procedure takes 10 minutes and has a sensitivity of 2ng BSA, higher than silver stains. Destain the membrane by simply rinsing in warm water for 10 minutes.
Staining and destaining does not affect the biological properties of the proteins. After destaining, protein bands can be probed with Western blot protocols and other analyses including sequencing work.
BLOT-FastStain™ is fully compatable with the LiCor Odyssey detection system and has succesfully been used as a loading control in this Odyssey system.
Application: Staining of membranes for visualization of proteins onto the membrane after Western transfer and dot-blot applications