Nunc® Barcode Scanners | Avantor
Nunc® Barcode Scanners
Nunc® Barcode Scanners
Certificates
Nunc® Barcode Scanners
Nunc® Barcode Scanners
1 Options Available Below
Product Details & Documents
Nunc bar code scanners with plug and play functionality play a critical role in sample tracking
The scanners automatically switch between code 128, code 39, interleave 2 of 5 and several other barcodes. These scanners are durable low-cost, and easy to use.
Product Family Options
Informations sur le produit
  • Availability
    Price
  • Discontinued
    Supplier Number: 330050
  • Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $1,870.47
    $1,870.47
    Case of 1
       $1,870.47
    Supplier Number: 330050