Cubitainer Containers, Low-Density Polyethylene, Qorpak® | Avantor
Cubitainer Containers, Low-Density Polyethylene, Qorpak®
Supplier:   Qorpak
Cubitainer Containers, Low-Density Polyethylene, Qorpak®
Supplier:  Qorpak
Certificates
Cubitainer Containers, Low-Density Polyethylene, Qorpak®
Cubitainer Containers, Low-Density Polyethylene, Qorpak®
1 Options Available Below
Product Details & Documents
Cubitainers® are low-density flexible polyethylene cube-shaped inserts inside a heavy-duty corrugated outer carton (32 ounce does not include carton).
  • Easy to ship, handle and pour
Designed to collapse as product is dispensed. The non-glug design assures product flow in a continuous, uninterrupted stream. A reversible well design permits opening to be raised or depressed for pouring and storage. Built to withstand the rigors of hazardous chemical packaging.
Product Family Options
Informations sur le produit
  • Capacité
    Color
    Taille du bouchon
    Cap Type
    Availability
    Price
  • 4 L (1 gal.)
    Natural
    38-400
    Discontinued
    Supplier Number: -
  • 4 L (1 gal.)
    Natural
    38-400
    White PP cap F217 lined
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 8
    Case of 8 - $430.46
    $430.46
    Prix promotionnel pour les nouveaux clients
    Case of 8
       $430.46
    Supplier Number: PLC-03458
Accessories
Informations sur le produit
  • Catalog #
    Material
    Color
    Availability
    Price
  • Quick Serve Tap for Cubitainers®
    Quick Serve Tap for Cubitainers®
    24301-704
    HDPE
    White
    Log in pour la disponibilité
    Each - $14.08