Individual Precision Analytical Weights, Class 3, Troemner
Individual Precision Analytical Weights, Class 3, Troemner
Supplier:  Troemner
Certificates
Individual Precision Analytical Weights, Class 3, Troemner
Individual Precision Analytical Weights, Class 3, Troemner
76 Options Available Below
Product Details & Documents
Highly polished kilogram and gram weights made of Troemner Alloy 8, which is a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage
Fractional weights 500 mg to 5 mg are made of stainless steel (30 mg weights are aluminum); fractional weights 3 mg and below are made of aluminum.
This exclusive material has a density of 8.0 at 25 °C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to –195.5 °C (–320 °F).
 : Standard weights supplied with a Statement of Accuracy. Where applicable, weights are supplied with a NIST/NVLAP Weight Calibration Certificate or a Traceable Certificate.
 : VWR Catalyst™ Services provides convenient and fast re-calibration services within 6 business days for your weights. Please contact us at 1.888.793.2300 or VWR[email protected] for a quote or more information. If your weights are found to be out of tolerance, we will make the proper adjustments to bring them back into tolerance. To prevent loss and ensure the timely processing and integrity of your precision weights and/or calibration devices, please do not pack in foam peanuts. Please avoid packing your items with foam, packing peanuts or other material that cannot be recycled.
Product Family Options
Informations sur le produit
  • Certification
    Weight
    Availability
    Price
  • Statement Of Accuracy
    2 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $201.18
    $201.18
    Prix promotionnel pour les nouveaux clients
    Each
       $201.18
    Supplier Number: 80858289
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12365-326
    Product available on GSA Advantage®
  • Statement Of Accuracy
    3 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $138.72
    $138.72
    Prix promotionnel pour les nouveaux clients
    Each
       $138.72
    Supplier Number: 80858275
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12365-328
    Product available on GSA Advantage®
  • Statement Of Accuracy
    5 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $143.84
    $143.84
    Prix promotionnel pour les nouveaux clients
    Each
       $143.84
    Supplier Number: 80858260
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12365-330
    Product available on GSA Advantage®
  • Statement Of Accuracy
    20 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $175.69
    $175.69
    Prix promotionnel pour les nouveaux clients
    Each
       $175.69
    Supplier Number: 80858229
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12365-334
    Product available on GSA Advantage®
  • Statement Of Accuracy
    30 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $181.45
    $181.45
    Prix promotionnel pour les nouveaux clients
    Each
       $181.45
    Supplier Number: 80858215
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12365-336
    Product available on GSA Advantage®
  • Statement Of Accuracy
    50 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $200.36
    $200.36
    Prix promotionnel pour les nouveaux clients
    Each
       $200.36
    Supplier Number: 80858200
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12365-338
    Product available on GSA Advantage®
  • Statement Of Accuracy
    100 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $205.38
    $205.38
    Prix promotionnel pour les nouveaux clients
    Each
       $205.38
    Supplier Number: 80858184
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12365-340
    Product available on GSA Advantage®
  • Statement Of Accuracy
    1 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $149.13
    $149.13
    Prix promotionnel pour les nouveaux clients
    Each
       $149.13
    Supplier Number: 80858708
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-300
    Product available on GSA Advantage®
  • Statement Of Accuracy
    2 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $149.30
    $149.30
    Prix promotionnel pour les nouveaux clients
    Each
       $149.30
    Supplier Number: 80858669
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-302
    Product available on GSA Advantage®
  • Statement Of Accuracy
    3 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $104.00
    $104.00
    Prix promotionnel pour les nouveaux clients
    Each
       $104.00
    Supplier Number: 80858639
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-304
    Product available on GSA Advantage®
  • Statement Of Accuracy
    5 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $149.13
    $149.13
    Prix promotionnel pour les nouveaux clients
    Each
       $149.13
    Supplier Number: 80858616
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-306
    Product available on GSA Advantage®
  • Statement Of Accuracy
    10 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $104.00
    $104.00
    Prix promotionnel pour les nouveaux clients
    Each
       $104.00
    Supplier Number: 80858577
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-308
    Product available on GSA Advantage®
  • Statement Of Accuracy
    20 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $104.00
    $104.00
    Prix promotionnel pour les nouveaux clients
    Each
       $104.00
    Supplier Number: 80858537
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-310
    Product available on GSA Advantage®
  • Statement Of Accuracy
    30 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $149.13
    $149.13
    Prix promotionnel pour les nouveaux clients
    Each
       $149.13
    Supplier Number: 80858507
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-312
    Product available on GSA Advantage®
  • Statement Of Accuracy
    50 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $104.00
    $104.00
    Prix promotionnel pour les nouveaux clients
    Each
       $104.00
    Supplier Number: 80858486
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-314
    Product available on GSA Advantage®
  • Statement Of Accuracy
    100 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $104.00
    $104.00
    Prix promotionnel pour les nouveaux clients
    Each
       $104.00
    Supplier Number: 80858448
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-316
    Product available on GSA Advantage®
  • Statement Of Accuracy
    200 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $104.00
    $104.00
    Prix promotionnel pour les nouveaux clients
    Each
       $104.00
    Supplier Number: 80858409
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-318
    Product available on GSA Advantage®
  • Statement Of Accuracy
    300 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $151.05
    $151.05
    Prix promotionnel pour les nouveaux clients
    Each
       $151.05
    Supplier Number: 80858374
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-320
    Product available on GSA Advantage®
  • Statement Of Accuracy
    500 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $104.00
    $104.00
    Prix promotionnel pour les nouveaux clients
    Each
       $104.00
    Supplier Number: 80858334
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-322
    Product available on GSA Advantage®
  • Statement Of Accuracy
    1 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $193.42
    $193.42
    Prix promotionnel pour les nouveaux clients
    Each
       $193.42
    Supplier Number: 80858304
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-324
    Product available on GSA Advantage®
  • NVLAP
    1 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $333.15
    $333.15
    Prix promotionnel pour les nouveaux clients
    Each
       $333.15
    Supplier Number: 80858711
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-345
    Product available on GSA Advantage®
  • NVLAP
    2 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $333.15
    $333.15
    Prix promotionnel pour les nouveaux clients
    Each
       $333.15
    Supplier Number: 80858672
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-346
    Product available on GSA Advantage®
  • NVLAP
    3 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $229.38
    $229.38
    Prix promotionnel pour les nouveaux clients
    Each
       $229.38
    Supplier Number: 80858641
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-347
    Product available on GSA Advantage®
  • NVLAP
    5 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $328.82
    $328.82
    Prix promotionnel pour les nouveaux clients
    Each
       $328.82
    Supplier Number: 80858619
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-348
    Product available on GSA Advantage®
  • NVLAP
    10 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $229.38
    $229.38
    Prix promotionnel pour les nouveaux clients
    Each
       $229.38
    Supplier Number: 80858580
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-349
    Product available on GSA Advantage®
  • NVLAP
    20 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $229.38
    $229.38
    Prix promotionnel pour les nouveaux clients
    Each
       $229.38
    Supplier Number: 80858540
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-350
    Product available on GSA Advantage®
  • NVLAP
    30 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $229.38
    $229.38
    Prix promotionnel pour les nouveaux clients
    Each
       $229.38
    Supplier Number: 80858509
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-351
    Product available on GSA Advantage®
  • NVLAP
    50 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $328.82
    $328.82
    Prix promotionnel pour les nouveaux clients
    Each
       $328.82
    Supplier Number: 80858489
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-352
    Product available on GSA Advantage®
  • NVLAP
    100 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $328.82
    $328.82
    Prix promotionnel pour les nouveaux clients
    Each
       $328.82
    Supplier Number: 80858451
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-353
    Product available on GSA Advantage®
  • NVLAP
    200 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $229.38
    $229.38
    Prix promotionnel pour les nouveaux clients
    Each
       $229.38
    Supplier Number: 80858412
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-354
    Product available on GSA Advantage®
  • NVLAP
    300 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $229.38
    $229.38
    Prix promotionnel pour les nouveaux clients
    Each
       $229.38
    Supplier Number: 80858377
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-355
    Product available on GSA Advantage®
  • NVLAP
    500 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $229.38
    $229.38
    Prix promotionnel pour les nouveaux clients
    Each
       $229.38
    Supplier Number: 80858337
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-356
    Product available on GSA Advantage®
  • NVLAP
    1 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $378.29
    $378.29
    Prix promotionnel pour les nouveaux clients
    Each
       $378.29
    Supplier Number: 80858306
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-357
    Product available on GSA Advantage®
  • NVLAP
    2 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $393.56
    $393.56
    Prix promotionnel pour les nouveaux clients
    Each
       $393.56
    Supplier Number: 80858291
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-358
    Product available on GSA Advantage®
  • NVLAP
    3 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $272.29
    $272.29
    Prix promotionnel pour les nouveaux clients
    Each
       $272.29
    Supplier Number: 80858277
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-359
    Product available on GSA Advantage®
  • NVLAP
    5 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $406.67
    $406.67
    Prix promotionnel pour les nouveaux clients
    Each
       $406.67
    Supplier Number: 80858262
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-360
    Product available on GSA Advantage®
  • NVLAP
    10 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $417.90
    $417.90
    Prix promotionnel pour les nouveaux clients
    Each
       $417.90
    Supplier Number: 80858246
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-361
    Product available on GSA Advantage®
  • NVLAP
    20 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $429.79
    $429.79
    Prix promotionnel pour les nouveaux clients
    Each
       $429.79
    Supplier Number: 80858232
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-362
    Product available on GSA Advantage®
  • NVLAP
    30 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $432.14
    $432.14
    Prix promotionnel pour les nouveaux clients
    Each
       $432.14
    Supplier Number: 80858217
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-363
    Product available on GSA Advantage®
  • NVLAP
    50 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $311.94
    $311.94
    Prix promotionnel pour les nouveaux clients
    Each
       $311.94
    Supplier Number: 80858203
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-364
    Product available on GSA Advantage®
  • NVLAP
    100 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $469.78
    $469.78
    Prix promotionnel pour les nouveaux clients
    Each
       $469.78
    Supplier Number: 80858187
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-365
    Product available on GSA Advantage®
  • Traceable
    1 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $175.94
    $175.94
    Prix promotionnel pour les nouveaux clients
    Each
       $175.94
    Supplier Number: 80858709
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-370
    Product available on GSA Advantage®
  • Traceable
    2 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $175.94
    $175.94
    Prix promotionnel pour les nouveaux clients
    Each
       $175.94
    Supplier Number: 80858670
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-371
    Product available on GSA Advantage®
  • Traceable
    3 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $175.94
    $175.94
    Prix promotionnel pour les nouveaux clients
    Each
       $175.94
    Supplier Number: 80858640
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-372
    Product available on GSA Advantage®
  • Traceable
    5 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $175.94
    $175.94
    Prix promotionnel pour les nouveaux clients
    Each
       $175.94
    Supplier Number: 80858617
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-373
    Product available on GSA Advantage®
  • Traceable
    10 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $252.55
    $252.55
    Prix promotionnel pour les nouveaux clients
    Each
       $252.55
    Supplier Number: 80858578
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-374
    Product available on GSA Advantage®
  • Traceable
    20 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $175.94
    $175.94
    Prix promotionnel pour les nouveaux clients
    Each
       $175.94
    Supplier Number: 80858538
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-375
    Product available on GSA Advantage®
  • Traceable
    30 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $175.94
    $175.94
    Prix promotionnel pour les nouveaux clients
    Each
       $175.94
    Supplier Number: 80858508
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-376
    Product available on GSA Advantage®
  • Traceable
    50 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $175.94
    $175.94
    Prix promotionnel pour les nouveaux clients
    Each
       $175.94
    Supplier Number: 80858487
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-377
    Product available on GSA Advantage®
  • Traceable
    100 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $251.98
    $251.98
    Prix promotionnel pour les nouveaux clients
    Each
       $251.98
    Supplier Number: 80858449
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-378
    Product available on GSA Advantage®
  • Traceable
    200 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $175.94
    $175.94
    Prix promotionnel pour les nouveaux clients
    Each
       $175.94
    Supplier Number: 80858410
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-379
    Product available on GSA Advantage®
  • Traceable
    300 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $175.94
    $175.94
    Prix promotionnel pour les nouveaux clients
    Each
       $175.94
    Supplier Number: 80858375
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-380
    Product available on GSA Advantage®
  • Traceable
    500 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $255.38
    $255.38
    Prix promotionnel pour les nouveaux clients
    Each
       $255.38
    Supplier Number: 80858335
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-381
    Product available on GSA Advantage®
  • Traceable
    2 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $210.82
    $210.82
    Prix promotionnel pour les nouveaux clients
    Each
       $210.82
    Supplier Number: 80858290
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-383
    Product available on GSA Advantage®
  • Traceable
    3 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $210.82
    $210.82
    Prix promotionnel pour les nouveaux clients
    Each
       $210.82
    Supplier Number: 80858276
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-384
    Product available on GSA Advantage®
  • Traceable
    5 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $221.54
    $221.54
    Prix promotionnel pour les nouveaux clients
    Each
       $221.54
    Supplier Number: 80858261
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-385
    Product available on GSA Advantage®
  • Traceable
    20 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $242.92
    $242.92
    Prix promotionnel pour les nouveaux clients
    Each
       $242.92
    Supplier Number: 80858231
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-387
    Product available on GSA Advantage®
  • Traceable
    30 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $260.90
    $260.90
    Prix promotionnel pour les nouveaux clients
    Each
       $260.90
    Supplier Number: 80858216
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-388
    Product available on GSA Advantage®
  • Traceable
    50 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $264.34
    $264.34
    Prix promotionnel pour les nouveaux clients
    Each
       $264.34
    Supplier Number: 80858202
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-389
    Product available on GSA Advantage®
  • Traceable
    100 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $276.99
    $276.99
    Prix promotionnel pour les nouveaux clients
    Each
       $276.99
    Supplier Number: 80858186
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-390
    Product available on GSA Advantage®
  • Statement Of Accuracy
    200 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $454.04
    $454.04
    Prix promotionnel pour les nouveaux clients
    Each
       $454.04
    Supplier Number: 80858168
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    14216-754
    Product available on GSA Advantage®
  • Traceable
    200 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $575.17
    $575.17
    Prix promotionnel pour les nouveaux clients
    Each
       $575.17
    Supplier Number: 80858169
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    14216-756
    Product available on GSA Advantage®
  • Statement Of Accuracy
    500 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $482.35
    $482.35
    Prix promotionnel pour les nouveaux clients
    Each
       $482.35
    Supplier Number: 80858141
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    14216-766
    Product available on GSA Advantage®
  • Traceable
    500 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $869.48
    $869.48
    Prix promotionnel pour les nouveaux clients
    Each
       $869.48
    Supplier Number: 80858142
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    14216-768
    Product available on GSA Advantage®
  • NVLAP
    500 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $961.38
    $961.38
    Prix promotionnel pour les nouveaux clients
    Each
       $961.38
    Supplier Number: 80858143
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    14216-770
    Product available on GSA Advantage®
  • Statement Of Accuracy
    1 kg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,031.49
    $1,031.49
    Prix promotionnel pour les nouveaux clients
    Each
       $1,031.49
    Supplier Number: 80858127
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    14216-778
    Product available on GSA Advantage®
  • Traceable
    1 kg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,675.94
    $1,675.94
    Prix promotionnel pour les nouveaux clients
    Each
       $1,675.94
    Supplier Number: 80858128
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    14216-780
    Product available on GSA Advantage®
  • NVLAP
    1 kg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,819.48
    $1,819.48
    Prix promotionnel pour les nouveaux clients
    Each
       $1,819.48
    Supplier Number: 80858129
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    14216-782
    Product available on GSA Advantage®
  • Statement Of Accuracy
    2 kg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,620.35
    $1,620.35
    Prix promotionnel pour les nouveaux clients
    Each
       $1,620.35
    Supplier Number: 80858114
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    14216-790
    Product available on GSA Advantage®
  • Traceable
    2 kg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,853.30
    $1,853.30
    Prix promotionnel pour les nouveaux clients
    Each
       $1,853.30
    Supplier Number: 80858115
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    14216-792
    Product available on GSA Advantage®
  • Statement Of Accuracy
    5 kg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,498.10
    $2,498.10
    Prix promotionnel pour les nouveaux clients
    Each
       $2,498.10
    Supplier Number: 80858087
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    14216-802
    Product available on GSA Advantage®
  • Traceable
    5 kg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,674.94
    $2,674.94
    Prix promotionnel pour les nouveaux clients
    Each
       $2,674.94
    Supplier Number: 80858088
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    14216-804
    Product available on GSA Advantage®
  • NVLAP
    5 kg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,959.02
    $2,959.02
    Prix promotionnel pour les nouveaux clients
    Each
       $2,959.02
    Supplier Number: 80858089
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    14216-806
    Product available on GSA Advantage®
  • Statement Of Accuracy
    10 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $241.65
    $241.65
    Prix promotionnel pour les nouveaux clients
    Each
       $241.65
    Supplier Number: 80858243
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12365-332
    Product available on GSA Advantage®
  • Traceable
    1 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $293.47
    $293.47
    Prix promotionnel pour les nouveaux clients
    Each
       $293.47
    Supplier Number: 80858305
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-382
    Product available on GSA Advantage®
  • Traceable
    10 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $242.92
    $242.92
    Prix promotionnel pour les nouveaux clients
    Each
       $242.92
    Supplier Number: 80858245
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    12765-386
    Product available on GSA Advantage®