Individual Precision Analytical Weights, Class 2, Troemner | Avantor
Individual Precision Analytical Weights, Class 2, Troemner
Individual Precision Analytical Weights, Class 2, Troemner
Supplier:  Troemner
Certificates
Individual Precision Analytical Weights, Class 2, Troemner
Individual Precision Analytical Weights, Class 2, Troemner
76 Options Available Below
Product Details & Documents
Highly polished kilogram and gram weights made of Troemner Alloy 8, which is a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage
Fractional weights 500 mg to 5 mg are made of stainless steel (30 mg weights are aluminum); fractional weights 3 mg and below are made of aluminum.
This exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to –195.5°C (–320°F).
Certifications: Where applicable, weights are supplied with a NIST/NVLAP Weight Calibration Certificate or a Traceable Certificate.
Services: VWR Catalyst™ Services provides convenient and fast re-calibration services within 6 business days for your weights. Please contact us at 1.888.793.2300 or VWR[email protected] for a quote or more information. If your weights are found to be out of tolerance, we will make the proper adjustments to bring them back into tolerance. To prevent loss and ensure the timely processing and integrity of your precision weights and/or calibration devices, please do not pack in foam peanuts. Please avoid packing your items with foam, packing peanuts or other material that cannot be recycled.
Product Family Options
Informations sur le produit
  • Certification
    Weight
    Availability
    Price
  • NVLAP
    1 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $245.81
    $245.81
    Prix promotionnel pour les nouveaux clients
    Each
       $245.81
    Supplier Number: 80858705
  • NVLAP
    2 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $245.81
    $245.81
    Prix promotionnel pour les nouveaux clients
    Each
       $245.81
    Supplier Number: 80858666
  • NVLAP
    3 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $245.81
    $245.81
    Prix promotionnel pour les nouveaux clients
    Each
       $245.81
    Supplier Number: 80858638
  • NVLAP
    5 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $354.61
    $354.61
    Prix promotionnel pour les nouveaux clients
    Each
       $354.61
    Supplier Number: 80858613
  • NVLAP
    10 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $356.28
    $356.28
    Prix promotionnel pour les nouveaux clients
    Each
       $356.28
    Supplier Number: 80858574
  • NVLAP
    20 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $356.28
    $356.28
    Prix promotionnel pour les nouveaux clients
    Each
       $356.28
    Supplier Number: 80858534
  • NVLAP
    30 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $245.81
    $245.81
    Prix promotionnel pour les nouveaux clients
    Each
       $245.81
    Supplier Number: 80858506
  • NVLAP
    50 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $356.28
    $356.28
    Prix promotionnel pour les nouveaux clients
    Each
       $356.28
    Supplier Number: 80858483
  • NVLAP
    100 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $356.18
    $356.18
    Prix promotionnel pour les nouveaux clients
    Each
       $356.18
    Supplier Number: 80858445
  • NVLAP
    200 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $356.28
    $356.28
    Prix promotionnel pour les nouveaux clients
    Each
       $356.28
    Supplier Number: 80858406
  • NVLAP
    300 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $245.38
    $245.38
    Prix promotionnel pour les nouveaux clients
    Each
       $245.38
    Supplier Number: 80858371
  • NVLAP
    500 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $356.18
    $356.18
    Prix promotionnel pour les nouveaux clients
    Each
       $356.18
    Supplier Number: 80858331
  • NVLAP
    1 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $390.78
    $390.78
    Prix promotionnel pour les nouveaux clients
    Each
       $390.78
    La quantité minimale de commande est de 1
    Supplier Number: 80858303
  • NVLAP
    2 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $288.11
    $288.11
    Prix promotionnel pour les nouveaux clients
    Each
       $288.11
    La quantité minimale de commande est de 1
    Supplier Number: 80858288
  • NVLAP
    3 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $287.86
    $287.86
    Prix promotionnel pour les nouveaux clients
    Each
       $287.86
    Supplier Number: 80858274
  • NVLAP
    5 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $436.97
    $436.97
    Prix promotionnel pour les nouveaux clients
    Each
       $436.97
    La quantité minimale de commande est de 1
    Supplier Number: 80858259
  • NVLAP
    10 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $446.79
    $446.79
    Prix promotionnel pour les nouveaux clients
    Each
       $446.79
    La quantité minimale de commande est de 1
    Supplier Number: 80858242
  • NVLAP
    20 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $446.50
    $446.50
    Prix promotionnel pour les nouveaux clients
    Each
       $446.50
    La quantité minimale de commande est de 1
    Supplier Number: 80858228
  • NVLAP
    30 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $317.22
    $317.22
    Prix promotionnel pour les nouveaux clients
    Each
       $317.22
    Supplier Number: 80858214
  • NVLAP
    50 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $474.93
    $474.93
    Prix promotionnel pour les nouveaux clients
    Each
       $474.93
    Supplier Number: 80858199
  • NVLAP
    100 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $487.73
    $487.73
    Prix promotionnel pour les nouveaux clients
    Each
       $487.73
    Supplier Number: 80858183
  • NVLAP
    200 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $819.90
    $819.90
    Prix promotionnel pour les nouveaux clients
    Each
       $819.90
    Supplier Number: 80858167
  • NVLAP
    500 g
    Discontinued
    Supplier Number: -
  • NVLAP
    1 kg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,370.02
    $1,370.02
    Prix promotionnel pour les nouveaux clients
    Each
       $1,370.02
    Supplier Number: 80858126
  • NVLAP
    2 kg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,027.33
    $2,027.33
    Prix promotionnel pour les nouveaux clients
    Each
       $2,027.33
    Supplier Number: 80858113
  • NVLAP
    5 kg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,075.97
    $3,075.97
    Prix promotionnel pour les nouveaux clients
    Each
       $3,075.97
    Supplier Number: 80858086
  • Traceable
    1 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $290.41
    $290.41
    Prix promotionnel pour les nouveaux clients
    Each
       $290.41
    Supplier Number: 80858703
  • Traceable
    2 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $290.41
    $290.41
    Prix promotionnel pour les nouveaux clients
    Each
       $290.41
    Supplier Number: 80858664
  • Traceable
    3 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $202.67
    $202.67
    Prix promotionnel pour les nouveaux clients
    Each
       $202.67
    Supplier Number: 80858637
  • Traceable
    5 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $290.41
    $290.41
    Prix promotionnel pour les nouveaux clients
    Each
       $290.41
    Supplier Number: 80858611
  • Traceable
    10 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $202.67
    $202.67
    Prix promotionnel pour les nouveaux clients
    Each
       $202.67
    La quantité minimale de commande est de 1
    Supplier Number: 80858572
  • Traceable
    20 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $202.67
    $202.67
    Prix promotionnel pour les nouveaux clients
    Each
       $202.67
    Supplier Number: 80858532
  • Traceable
    30 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $202.67
    $202.67
    Prix promotionnel pour les nouveaux clients
    Each
       $202.67
    Supplier Number: 80858505
  • Traceable
    50 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $292.82
    $292.82
    Prix promotionnel pour les nouveaux clients
    Each
       $292.82
    Supplier Number: 80858481
  • Traceable
    100 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $294.22
    $294.22
    Prix promotionnel pour les nouveaux clients
    Each
       $294.22
    Supplier Number: 80858443
  • Traceable
    200 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $202.67
    $202.67
    Prix promotionnel pour les nouveaux clients
    Each
       $202.67
    Supplier Number: 80858404
  • Traceable
    300 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $202.67
    $202.67
    Prix promotionnel pour les nouveaux clients
    Each
       $202.67
    Supplier Number: 80858369
  • Traceable
    500 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $290.41
    $290.41
    Prix promotionnel pour les nouveaux clients
    Each
       $290.41
    Supplier Number: 80858329
  • Traceable
    1 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $229.38
    $229.38
    Prix promotionnel pour les nouveaux clients
    Each
       $229.38
    La quantité minimale de commande est de 1
    Supplier Number: 80858302
  • Traceable
    2 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $351.36
    $351.36
    Prix promotionnel pour les nouveaux clients
    Each
       $351.36
    Supplier Number: 80858287
  • Traceable
    3 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $245.22
    $245.22
    Prix promotionnel pour les nouveaux clients
    Each
       $245.22
    Supplier Number: 80858273
  • Traceable
    5 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $379.32
    $379.32
    Prix promotionnel pour les nouveaux clients
    Each
       $379.32
    Supplier Number: 80858258
  • Traceable
    10 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $394.48
    $394.48
    Prix promotionnel pour les nouveaux clients
    Each
       $394.48
    Supplier Number: 80858241
  • Traceable
    20 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $389.31
    $389.31
    Prix promotionnel pour les nouveaux clients
    Each
       $389.31
    Supplier Number: 80858227
  • Traceable
    30 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $415.33
    $415.33
    Prix promotionnel pour les nouveaux clients
    Each
       $415.33
    Supplier Number: 80858213
  • Traceable
    50 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $419.86
    $419.86
    Prix promotionnel pour les nouveaux clients
    Each
       $419.86
    Supplier Number: 80858198
  • Traceable
    100 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $306.67
    $306.67
    Prix promotionnel pour les nouveaux clients
    Each
       $306.67
    La quantité minimale de commande est de 1
    Supplier Number: 80858182
  • Traceable
    200 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $648.83
    $648.83
    Prix promotionnel pour les nouveaux clients
    Each
       $648.83
    La quantité minimale de commande est de 1
    Supplier Number: 80858166
  • Traceable
    500 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $862.18
    $862.18
    Prix promotionnel pour les nouveaux clients
    Each
       $862.18
    Supplier Number: 80858139
  • Traceable
    1 kg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,773.62
    $1,773.62
    Prix promotionnel pour les nouveaux clients
    Each
       $1,773.62
    Supplier Number: 80858125
  • Traceable
    2 kg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,900.30
    $1,900.30
    Prix promotionnel pour les nouveaux clients
    Each
       $1,900.30
    Supplier Number: 80858112
  • Traceable
    5 kg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,774.92
    $2,774.92
    Prix promotionnel pour les nouveaux clients
    Each
       $2,774.92
    Supplier Number: 80858085
  • Without certification
    1 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $136.10
    $136.10
    Prix promotionnel pour les nouveaux clients
    Each
       $136.10
    Supplier Number: 80858701
  • Without certification
    2 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $136.10
    $136.10
    Prix promotionnel pour les nouveaux clients
    Each
       $136.10
    Supplier Number: 80858663
  • Without certification
    3 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $136.10
    $136.10
    Prix promotionnel pour les nouveaux clients
    Each
       $136.10
    Supplier Number: 80858636
  • Without certification
    5 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $136.10
    $136.10
    Prix promotionnel pour les nouveaux clients
    Each
       $136.10
    Supplier Number: 80858610
  • Without certification
    10 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $136.10
    $136.10
    Prix promotionnel pour les nouveaux clients
    Each
       $136.10
    Supplier Number: 80858569
  • Without certification
    20 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $136.10
    $136.10
    Prix promotionnel pour les nouveaux clients
    Each
       $136.10
    Supplier Number: 80858530
  • Without certification
    30 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $136.10
    $136.10
    Prix promotionnel pour les nouveaux clients
    Each
       $136.10
    Supplier Number: 80858504
  • Without certification
    50 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $136.10
    $136.10
    Prix promotionnel pour les nouveaux clients
    Each
       $136.10
    Supplier Number: 80858480
  • Without certification
    100 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $136.10
    $136.10
    Prix promotionnel pour les nouveaux clients
    Each
       $136.10
    Supplier Number: 80858442
  • Without certification
    200 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $196.46
    $196.46
    Prix promotionnel pour les nouveaux clients
    Each
       $196.46
    Supplier Number: 80858403
  • Without certification
    300 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $136.10
    $136.10
    Prix promotionnel pour les nouveaux clients
    Each
       $136.10
    Supplier Number: 80858368
  • Without certification
    500 mg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $194.79
    $194.79
    Prix promotionnel pour les nouveaux clients
    Each
       $194.79
    Supplier Number: 80858328
  • Without certification
    1 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $151.86
    $151.86
    Prix promotionnel pour les nouveaux clients
    Each
       $151.86
    Supplier Number: 80858301
  • Without certification
    2 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $232.62
    $232.62
    Prix promotionnel pour les nouveaux clients
    Each
       $232.62
    Supplier Number: 80858286
  • Without certification
    3 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $160.52
    $160.52
    Prix promotionnel pour les nouveaux clients
    Each
       $160.52
    Supplier Number: 80858272
  • Without certification
    5 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $173.28
    $173.28
    Prix promotionnel pour les nouveaux clients
    Each
       $173.28
    Supplier Number: 80858256
  • Without certification
    10 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $271.12
    $271.12
    Prix promotionnel pour les nouveaux clients
    Each
       $271.12
    Supplier Number: 80858240
  • Without certification
    20 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $189.30
    $189.30
    Prix promotionnel pour les nouveaux clients
    Each
       $189.30
    Supplier Number: 80858226
  • Without certification
    30 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $197.10
    $197.10
    Prix promotionnel pour les nouveaux clients
    Each
       $197.10
    Supplier Number: 80858212
  • Without certification
    50 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $309.62
    $309.62
    Prix promotionnel pour les nouveaux clients
    Each
       $309.62
    Supplier Number: 80858197
  • Without certification
    100 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $224.01
    $224.01
    Prix promotionnel pour les nouveaux clients
    Each
       $224.01
    Supplier Number: 80858181
  • Without certification
    500 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $504.12
    $504.12
    Prix promotionnel pour les nouveaux clients
    Each
       $504.12
    Supplier Number: 80858138
  • Without certification
    1 kg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,074.22
    $1,074.22
    Prix promotionnel pour les nouveaux clients
    Each
       $1,074.22
    Supplier Number: 80858124
  • Without certification
    5 kg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,572.34
    $2,572.34
    Prix promotionnel pour les nouveaux clients
    Each
       $2,572.34
    Supplier Number: 80858084
  • Without certification
    200 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $510.13
    $510.13
    Prix promotionnel pour les nouveaux clients
    Each
       $510.13
    Supplier Number: 80858165