Nunc® Multidishes, Non-treated, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
These multi dishes are non-treated and can be used for the suspension of cell cultures
The dishes have raised well rims which lowers the risk of cross-contamination. They also have flat bottom wells that allow for easy viewing of cultures. Lids with notched corners for a good fit are included. The lids help with single position orientation and cross contamination prevention. The multi dishes are made of sterile polystyrene.
Frequently Bought Together