ContactUsLinkComponent
Frog Liver GenLysate™ Frog Lysate, G-Biosciences
Frog Liver GenLysate™ Frog Lysate, G-Biosciences
Catalog # CA82021-672
Supplier:  G-Biosciences
CAS Number:  
Frog Liver GenLysate™ Frog Lysate, G-Biosciences
Catalog # CA82021-672
Supplier:  G-Biosciences
Supplier Number:  ML-04
CAS Number:  
Specifications
  • Cell Format:
    Cryopreserved
  • Description:
    Normal Tissue, Frog Liver GenLysate™
  • Size:
    150 µg
  • Cat. no.:
    CA82021-672
  • Source:
    Frog
  • Tissue:
    Liver
  • Supplier No.:
    ML-04
Specifications
About this item
G-Biosciences' GenLysates™ are total protein tissue and cell lysates that are extracted from the tissue or cell of interest. To inhibit proteolysis, the GenLysates™ are prepared in a lysis buffer containing a complete protease inhibitor cocktail. Each lysate lot is tested by electrophoresis and Western blot analysis.
G-Biosciences' GenLysate™ total protein lysates are ready to use for SDS-PAGE and Western blotting purposes
G-Biosciences' GenLysate™ total protein lysates are offered as a 150 µg lyophilized protein per vial and, following reconstitution in DI water, are ready for use. G-Biosciences offers GenLysate™ in a wide variety of different species and tissues. Species include: human (both normal tissue and tumor tissue lysates available), mouse, rat, macaque (primate), rabbit, pig, plant-related, bacteria, insect, bird, frog, fish, marsupial, earthworm, and yeast.
The total proteins are being extracted from whole tissue or cells following the method of Laemmli. The proteins are extracted in a denaturing buffer containing 62 mM Tris·HCl and 2% SDS with G-Biosciences' ProteaseARREST™ protease inhibitor cocktail. This minimizes proteolytic damage to the proteins.