Optimizer Buffer™ For Optimal Cross-linking & Modification, G-Biosciences
Optimizer Buffer™ For Optimal Cross-linking & Modification, G-Biosciences
Supplier:   G-Biosciences
Optimizer Buffer™ For Optimal Cross-linking & Modification, G-Biosciences
Supplier:  G-Biosciences
Certificates
Optimizer Buffer™ For Optimal Cross-linking & Modification, G-Biosciences
Optimizer Buffer™ For Optimal Cross-linking & Modification, G-Biosciences
6 Options Available Below
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
The conjugation and modification reactions used to cross-link proteins or couple labels to proteins, such as biotin, enzymes, and fluorescent dyes, require certain conditions, including pH and chemical composition, for optimal conjugation
G-Biosciences has prepared six reaction-specific buffers that provide the optimal conditions for protein labeling, modification, and cross reaction. Optimizer Buffer™ -I is free of amines and is specific for amine reactive groups, such as imidoesters and N-hydroxysuccinimide (NHS) esters. Optimizer Buffer™-II has a high pH and is specifically designed for the sulfhydryl reactive haloacetyls, such as iodoacetyl. Optimizer Buffer™-III is designed for sulfhydryl reactive groups, maleimides and pyridyl disulfides, and has reagents that prevent oxidation of free sulfhydryls. Optimizer Buffer™-IV is designed to provide the optimal conditions necessary for carboxyl reactive reagents. Optimizer Buffer™-V is a modified acetate buffer that is specific for carbohydrate reactive groups. Optimizer Buffer™-VI is an alkaline buffer specific for the formation of Schiff’s base between amines and glyoxal groups.
G-Biosciences' buffers contain optimized concentrations of buffering agents, pH and other cofactors for specific reactions. Simply exchange the buffer of your sample with a suitable Optimizer Buffer™, and you are ready for efficient reaction. Use of G-Biosciences' SpinOUT™ or Tube-O-DIALYZER™ is recommended for buffer exchange and optimal reaction results. Each Optimizer Buffer™ is supplied as a 5X concentrated buffer.
Many common buffers routinely used in laboratories have an inhibitory effect on conjugation reactions; for example, Tris buffers inhibit coupling to amines.
Product Family Options
Informations sur le produit
  • Désignation
    Reactive Group
    Target Group
    Availability
    Price
  • Optimizer Buffer™-I [5X]
    NHS-ester & imidoester groups
    Amine & Photoreactive
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2 (25ml)
    Pack of 2 (25ml) - $551.53
    $551.53
    Pack of 2 (25ml)
       $551.53
    Minimum order quantity is 1
    Supplier Number: BKC-04
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Optimizer Buffer™-II [5X]
    Iodoacetyl groups
    Sulfhydryl
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2 (25ml)
    Pack of 2 (25ml) - $551.53
    $551.53
    Pack of 2 (25ml)
       $551.53
    Minimum order quantity is 1
    Supplier Number: BKC-05
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Optimizer Buffer™-III [5X]
    Maleimides & pyridyl sulfides
    Sulfhydryl
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2 (25ml)
    Pack of 2 (25ml) - $551.53
    $551.53
    Pack of 2 (25ml)
       $551.53
    Minimum order quantity is 1
    Supplier Number: BKC-06
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Optimizer Buffer™-IV [5X]
    Carbodiimides
    Carboxyl
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2 (25ml)
    Pack of 2 (25ml) - $551.36
    $551.36
    Pack of 2 (25ml)
       $551.36
    Minimum order quantity is 1
    Supplier Number: BKC-07
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Optimizer Buffer™-V [5X]
    Hydrazide groups
    Carbohydrate
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2 (25ml)
    Pack of 2 (25ml) - $551.53
    $551.53
    Pack of 2 (25ml)
       $551.53
    Minimum order quantity is 1
    Supplier Number: BKC-08
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Optimizer Buffer™-VI [5X]
    Glyoxal groups
    Amine
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2 (25ml)
    Pack of 2 (25ml) - $551.53
    $551.53
    Pack of 2 (25ml)
       $551.53
    Minimum order quantity is 1
    Supplier Number: BKC-09
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.