ContactUsLinkComponent
VWR® Circulating Open Bath Systems, 240 V
Fournisseur:  VWR International
Certificats
undefined
VWR® Circulating Open Bath Systems, 240 V
Fournisseur:  VWR International
Spécifications
  • Catalog No:
  • 89202-916
  • 89202-912
  • Max. pressure:
  • 1.8 psi (0.12 bar)
  • 1.8 psi (0.12 bar)
  • Max. pressure flow rate:
  • 3.1 gpm (11.9 l/min.)
  • 3.1 gpm (11.9 l/min.)
  • Temperature stability:
  • ±0.07°C
  • ±0.07°C
  • Electrical:
  • 240 V, 50 Hz
  • 240 V, 50 Hz