Nalgene® IP2 High Density Polyethylene Wide Mouth Bottles, Bulk Pack, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Nalgene® IP2 high density polyethylene wide-mouth bottles are leak proof and meet ICAO and IATA requirements.
These bottles are safe, economical and are for customers designing, assembling and certifying combination packaging. The bottles are recommended for storage and shipment of liquid reagents.
Ordering information: Bottles and closures are bulk packed in separate polybags within the same carton.
Specifications
- Catalog No:
- 16059-726
- 16059-728
- 16059-730
- 16059-732
- Environmentally Preferable:
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)