ContactUsLinkComponent
NI™ (Non-Interfering™) Protein Assay, G-Biosciences
Catalogue # CA82022-356
Fournisseur:  G-Biosciences
Numéro CAS:  
undefined
NI™ (Non-Interfering™) Protein Assay, G-Biosciences
Catalogue # CA82022-356
Fournisseur:  G-Biosciences
Supplier Number:  786-005
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    NI™ (Non-Interfering™) Protein Assay kit with Albumin Standard
  • Size:
    500 Preps
  • Cat. no.:
    CA82022-356
  • Supplier No.:
    786-005
Spécifications
À propos de cet article
A highly sensitive colorimetric protein assay that overcomes interference of common laboratory agents present in protein solutions and shows minimal protein-to-protein variation
Universal Protein Precipitating Agent (UPPA™) is added to the protein solutions to rapidly precipitate total protein. Protein is immobilized by centrifugation and interfering agents in the supernatant are discarded. Protein concentration is assayed by mixing with an alkaline copper solution; the copper ions bind to the peptide backbone and the assay measures the unbound copper ions. The assay is independent of protein side chains minimizing protein-to-protein variation (see graph). The color density is inversely proportional to the amount of protein.
Has a linear response of 0.5 µg - 50 µg protein while only requiring 1-50µL of sample. Total protocol time for this product is around 30 minutes.
The assay is unaffected by the presence of common laboratory agents such as reducing agents, chelating agents, detergents, amines, sugars, chaotropes, salts, drugs, antibiotics, cobalt, and other common laboratory items (see table). The NI™ Protein Assay is composed of two simple steps (see figure). A highly sensitive colorimetric protein assay that overcomes interference of common laboratory agents present in protein solutions and shows minimal protein-to-protein variation. The assay is unaffected by the presence of common laboratory agents such as reducing agents, chelating agents, detergents, amines, sugars, chaotropes, salts, drugs, antibiotics, cobalt, and other common laboratory items (see table). The NI™ Protein Assay is composed of two simple steps (see figure).