To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Stainless Steel Cleaner
- Size:470 mL (15.89 OZ)
- Environmentally Preferable:Safer Human and Environmental Health
- Cat. no.:89208-418
- Supplier No.:SC1-P
Specifications
About this item
Gel Stainless Steel Cleaner uses advanced no-drip technology to eliminate streaks and drips when applied to stainless, metal and other hard surfaces.
Spray gel clings on contact and glides over most surfaces with minimum effort. When dealing with rust, oxidation and residue build up, the gel can be applied to seams and crevices to pre-treat before wiping.
The gel is not atomized into ambient air and so will not impact indoor air quality. Contains no phosphorus and has biodegradable surfactants. No VOC's.