ContactUsLinkComponent
FOCUS™ Mitochondria, G-Biosciences
FOCUS™ Mitochondria, G-Biosciences
Catalog # CA82023-258
Supplier:  G-Biosciences
CAS Number:  
FOCUS™ Mitochondria, G-Biosciences
Catalog # CA82023-258
Supplier:  G-Biosciences
Supplier Number:  786-022
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    FOCUS™ Mitochondria Kit
  • Size:
    50 Preps
  • Cat. no.:
    CA82023-258
  • Supplier No.:
    786-022
Specifications
About this item
Specifically designed for the isolation of intact mitochondria from cultured mammalian cells
This kit is highly adaptable for use with animal tissues and other sources of mitochondria. The kit is designed for 50 preps, where one prep is equivalent to 2x10^7 mammalian cells or 100 mg mammalian tissue.
This kit allows for the fast and efficient fractionation of the cytoplasm of cultured mammalian cells into an enriched fraction of mitochondria. The majority (>90%) of the mitochondria have intact inner and outer membranes and therefore retain their functionality.
Ordering information: Accessories available.