ContactUsLinkComponent
FOCUS™ Cytoplasmic and Nuclear, G-Biosciences
Catalog # CA82022-674
Supplier:  G-Biosciences
CAS Number:  
FOCUS™ Cytoplasmic and Nuclear, G-Biosciences
Catalog # CA82022-674
Supplier:  G-Biosciences
Supplier Number:  786-248
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Cytoplasmic and Nuclear Protein Kit
  • Size:
    50 Preps
  • Cat. no.:
    CA82022-674
  • Supplier No.:
    786-248
Specifications
About this item
Supplied with a strong chaotropic extraction buffer to solubilize both cytoplasmic and nuclear proteins, which is fully compatible with 2D gel electrophoresis.
G-Biosciences' FOCUS™ Cytoplasmic & Nuclear proteins fractionation kit is based on the Nuclear & Cytoplasmic Extraction kit (see figure), however it has been modified to be fully compatible with 2D electrophoresis and subsequent downstream processes.
The kit is provided with reagents necessary for fractionation of cytoplasmic and nuclear proteins as well as solubilization buffer suitable for IEF/2D gels for better spot resolution.
Designed for >50 preps, depending on the tissue used, where one prep is 20x10^6 mammalian cells or 100 mg mammalian tissue.