ContactUsLinkComponent
Nalgene® PFA Beakers with Handles, Thermo Scientific
Catalogue # 13917-574
Fournisseur #: DS1511-1000
undefined
Nalgene® PFA Beakers with Handles, Thermo Scientific
Catalogue # 13917-574
Supplier Number:  DS1511-1000
Fournisseur #: DS1511-1000
S'identifier pour la disponibilité
Select an Option
946.3 ml (32 oz.)
Select an Option
13917-574
Select an Option
DS1511-1000
À propos de cet article
Nalgene® PFA Beakers with Handles feature extraordinary chemical resistance and can be used repeatedly for high-purity applications
  • Rigid and translucent, enabling easy and consistent viewing of liquid levels
  • Molded in graduations in both oz. and mL that won't wear off with continuous use or washings
  • PFA is "non-stick" making the beaker easy to clean and resistant to contamination
The beakers can be used (and re-used) with strong oxidizing agents, acids, hydrocarbons and ketones. They can withstand temperatures from -270°C to +250°C, but are not for use on hot plates.
Frequently Bought Together