Advanced TC™ Treated Cell Culture Dishes, Vented, Sterile, Greiner Bio-One
Advanced Tissue Culture (TC) treated Cell Culture Dishes, made from virgin polystyrene, allow for optimal cultivation conditions for sensitive cells and cells cultivated under restricted growth conditions.
- Virgin polystyrene construction
- Free of RNase, DNase, and human DNA
- Nonpyrogenic
- Sterile
Advanced TC™ surface treatments provide the conditions for improved cell adherence, accelerated cell expansion and increase overall cell yield.
Advanced TC™ Cell Culture Dishes can be used with serum-reduced or serum-free media, transfection, transduction and high througput analysis. Dishes are vented for optimal gas exchange.
Sterile, DNase- Free, RNase- Free, human DNA - Free, nonpyrogenic and noncytotoxic. Store at room temperature. Two year shelf life.
: Sterile and user friendly packaging. Available with clear lid.
Frequently Bought Together