ContactUsLinkComponent
InGel™ Silver for Protein Digestion, G-Biosciences
  CA82021-586
 :  
undefined
InGel™ Silver for Protein Digestion, G-Biosciences
  CA82021-586
 :  786-241
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    InGel™ Silver
  • Size:
    100 preps
  • Cat. no.:
    CA82021-586
  • Supplier No.:
    786-241
G-Biosciences' InGel™ Silver provides a complete set of reagents for the in gel tryptic digestion and extraction of peptides for mass spectrometry (MALDI and LC MS/MS)
The protein spots are first excised from the silver stained gel and transferred to a proteomic grade tube. Silver stained gel pieces are washed with SilverOUT™ to remove inhibitory silver ions. The protein is then alkylated and reduced within the gel piece using the supplied aliquots of OneQuant™ DTT and OneQuant™ Iodoacetamide. The proteins are then digested within the gel using our Mass Spectrometry Grade Trypsin and proprietary Digestion Buffer.
The digested peptides are extracted with Pep-Extract™, a high diffusion peptide extraction buffer. The extracted peptides are suitable for mass spectrometry analysis without any subsequent treatments or cleaning procedures.
G-Biosciences' InGel™ Silver kit is supplied with SilverOUT™ for the removal of silver ions, OneQuant™ DTT used as a reducing agent and supplied in single use aliquots to prevent contamination, OneQuant™ Iodoacetamide used as an alkylating agent, again in single use aliquots to prevent contamination, Mass Spectrometry Grade Trypsin, Trypsin Digestion Buffer, and Pep-Extract™ for high level peptide extraction.
The kit is specifically designed for use with silver stained protein spots/bands.