Well-Coated™ Glutathione for Protein/Peptide Binding, G-Biosciences
Supplier: G-Biosciences
Certificates
About this item
G-Biosciences' Well-Coated™ Glutathione plates are designed to specifically bind GST (Glutathione S-Transferase) tagged proteins and peptides.
G-Biosciences' Well-Coated™ Glutathione plates have immobilized glutathione and can be used for the isolation of GST-tagged proteins direct from bacterial lysates and then used for subsequent ELISA protocols.
The wells are coated to a 100µL depth and are supplied pre-blocked in our proprietary Superior™ Blocking Buffer. The clear, white and black plates are offered for colorimetric, chemiluminescence and fluorescent detection systems, respectively.
Ordering information: Accessories available.
Frequently Bought Together