Nunc CryoTube Colored Caps, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Nunc CryoTube colored caps are replacement polypropylene screw caps with a silicone gasket to provide the best seal
They are made to be used with Nunc internally threaded CryoTube vials.
Frequently Bought Together