OPTI-SOLV® Mini Filters
About this item
The OPTI-SOLV® Mini Filter provides low-impact filtration in a package no larger than a finger-tight fitting.
- Hand-tight - no tools required
- Rated to 6,000psi
- Designed to filter out particles to extend column life
- Perfect Zero-Dead-Volume connections every time
- Floating stem automatically adjusts to any depth port
Use the Mini Filter to extend the life of analytical columns or before mass spectrometers as a last line of defense against debris. Perfect zero-dead-volume connections are obtained every time with the patented automatic tube stop depth adjustment. Unique depth-filtering elements allow for greater capacity with no band spreading or loss of performance. The efficient design requires no holder and threads directly into any 10 to 32 port.
The OPTI-SOLV Mini, Micro and Nano filters are easy to use and perform exceptionally well for volume-critical applications.
Delivery information: Includes five disposable cartridges.
3 Options Available Below
Product Details & Documents
The OPTI-SOLV® Mini Filter provides low-impact filtration in a package no larger than a finger-tight fitting.
- Hand-tight - no tools required
- Rated to 6,000psi
- Designed to filter out particles to extend column life
- Perfect Zero-Dead-Volume connections every time
- Floating stem automatically adjusts to any depth port
Use the Mini Filter to extend the life of analytical columns or before mass spectrometers as a last line of defense against debris. Perfect zero-dead-volume connections are obtained every time with the patented automatic tube stop depth adjustment. Unique depth-filtering elements allow for greater capacity with no band spreading or loss of performance. The efficient design requires no holder and threads directly into any 10 to 32 port.
The OPTI-SOLV Mini, Micro and Nano filters are easy to use and perform exceptionally well for volume-critical applications.
Delivery information: Includes five disposable cartridges.
Product Family Options
Product Information
- DésignationTaille de poreAvailabilityPrice
- Supplier Number: 10-04-00095
- Supplier Number: 10-04-00097
- Supplier Number: 10-04-02768